Gene Transcripts

Gene Symbol
Ttc5
Entrez Gene ID
305837
Gene Name
tetratricopeptide repeat domain 5
Gene Aliases
Chromosome Location
Chr. 15: 27698375 - 27715970 on Build Rnor_6.0
UniGene ID
Rn.16620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013131.1 NP_001013149.1 5, 6 662
GenBank mRNA BC091207.1 - 5, 6 662

View Full Product Details