Gene Transcripts

Gene Symbol
Efr3b
Entrez Gene ID
313928
Gene Name
EFR3 homolog B (S. cerevisiae)
Gene Aliases
RGD1310845
Chromosome Location
Chr. 6: 28390541 - 28464174 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239853.1 XP_006239915.1 16, 17 1956

View Full Product Details