Gene Transcripts

Gene Symbol
Aox4
Entrez Gene ID
316424
Gene Name
aldehyde oxidase 4
Gene Aliases
Aa2-245, Aoh2, RGD1311975
Chromosome Location
Chr. 9: 65110330 - 65167188 on Build Rnor_6.0
UniGene ID
Rn.198845
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008523.1 NP_001008523.1 26 3019
XM_006244987.2 XP_006245049.1 26 2990
GenBank mRNA AY665587.1 - 26 3019

View Full Product Details