Gene Transcripts

Gene Symbol
MGC94207
Entrez Gene ID
362946
Gene Name
similar to RIKEN cDNA C030006K11
Gene Aliases
Chromosome Location
Chr. 7: 117786206 - 117788550 on Build Rnor_6.0
UniGene ID
Rn.140135
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007751.1 NP_001007752.1 3 553
GenBank mRNA BC081992.1 - 3 553

View Full Product Details