Gene Transcripts

Gene Symbol
Mid2
Entrez Gene ID
363502
Gene Name
midline 2
Gene Aliases
RGD1565674
Chromosome Location
Chr. X: 112019987 - 112121980 on Build Rnor_6.0
UniGene ID
Rn.15520
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191889.1 NP_001178818.1 2 251
XM_006234247.2 XP_006234309.1 2 636
XM_006234248.2 XP_006234310.1 2 192

View Full Product Details