Gene Transcripts

Gene Symbol
Sp3
Entrez Gene ID
367846
Gene Name
Sp3 transcription factor
Gene Aliases
Chromosome Location
Chr. 3: 59644359 - 59689011 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761975.1 XP_008760197.1 4 695
GenBank mRNA AA875516.1 - 2 197

View Full Product Details