Gene Transcripts

Gene Symbol
Olr1166
Entrez Gene ID
405164
Gene Name
olfactory receptor 1166
Gene Aliases
Chromosome Location
Chr. 8: 20265891 - 20266829 on Build Rnor_6.0
UniGene ID
Rn.143583
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000867.1 NP_001000867.1 1 102

Gene Symbol
LOC100911583
Entrez Gene ID
100911583
Gene Name
olfactory receptor 7E24-like
Gene Aliases
Chromosome Location
Chr. 8: 21073842 - 21082605 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750446.2 XP_003750494.2 2 135

Gene Symbol
LOC690764
Entrez Gene ID
690764
Gene Name
similar to olfactory receptor Olr1166
Gene Aliases
Chromosome Location
Chr. 8: 20480747 - 20481634 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766033.1 XP_008764255.1 51

View Full Product Details