Gene Transcripts

Gene Symbol
DGKI
Entrez Gene ID
9162
Gene Name
diacylglycerol kinase iota
Gene Aliases
DGK-IOTA
Chromosome Location
Chr. 7: 137384283 - 137846864 on Build GRCh38
UniGene ID
Hs.242947
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321710.1 NP_001308639.1 21, 22 2214
NM_004717.3 NP_004708.1 21, 22 2248
GenBank mRNA AB209167.1 - 21, 22 1886

View Full Product Details