Gene Transcripts

Gene Symbol
Btf3
Entrez Gene ID
294680
Gene Name
basic transcription factor 3
Gene Aliases
Chromosome Location
Chr. 2: 28417316 - 28424453 on Build Rnor_6.0
UniGene ID
Rn.1858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008309.1 NP_001008310.1 6 772
XM_006231835.2 XP_006231897.1 6 1042
GenBank mRNA BC085343.1 - 6 772
FQ229173.1 - 6 778

View Full Product Details