Gene Transcripts

Gene Symbol
Cdca8
Entrez Gene ID
500545
Gene Name
cell division cycle associated 8
Gene Aliases
Chromosome Location
Chr. 5: 142911310 - 142933536 on Build Rnor_6.0
UniGene ID
Rn.14970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025050.1 NP_001020221.1 4 443
XM_008764064.1 XP_008762286.1 5 443
GenBank mRNA BC079274.1 - 4 443
BC079293.1 - 5 422 7134482

View Full Product Details