Gene Transcripts

Gene Symbol
RGD1564854
Entrez Gene ID
502617
Gene Name
similar to divalent cation tolerant protein CUTA
Gene Aliases
Chromosome Location
Chr. 3: 13936715 - 13955651 on Build Rnor_6.0
UniGene ID
Rn.139718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109341.1 NP_001102811.1 4, 5 380
XM_006233991.2 XP_006234053.1 4, 5 448

View Full Product Details