Gene Transcripts

Gene Symbol
Tsfm
Entrez Gene ID
679068
Gene Name
Ts translation elongation factor, mitochondrial
Gene Aliases
2A3-2, EF-Ts, EF-TsMt
Chromosome Location
Chr. 7: 70311948 - 70319346 on Build Rnor_6.0
UniGene ID
Rn.216537
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276476.1 NP_001263405.1 6 591
GenBank mRNA AJ005161.1 - 6 592
FQ220263.1 - 6 591

View Full Product Details