Gene Transcripts

Gene Symbol
HS3ST6
Entrez Gene ID
64711
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 6
Gene Aliases
3-OST-6, HS3ST5
Chromosome Location
Chr. 16: 1911463 - 1921428 on Build GRCh38
UniGene ID
Hs.670144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009606.3 NP_001009606.3 2 814
XM_011522608.2 XP_011520910.1 2 1040
XM_011522609.2 XP_011520911.1 2 2521

View Full Product Details