Gene Transcripts

Gene Symbol
ZNF263
Entrez Gene ID
10127
Gene Name
zinc finger protein 263
Gene Aliases
FPM315, ZKSCAN12, ZSCAN44
Chromosome Location
Chr. 16: 3283183 - 3301401 on Build GRCh38
UniGene ID
Hs.611475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005741.4 NP_005732.2 6 1948
XM_006720831.1 XP_006720894.1 6 2016
XM_006720832.1 XP_006720895.1 5 1938
XM_006720833.1 XP_006720896.1 4 1800
XM_006720834.3 XP_006720897.1 5 2071
XM_011522344.1 XP_011520646.1 6 2055
XM_017022850.1 XP_016878339.1 5 1494
GenBank mRNA AK295350.1 - 2 814
AK312421.1 - 6 1930
BC008805.2 - 6 1946 3945548
D88827.1 - 6 1908

View Full Product Details