Gene Transcripts

Gene Symbol
Ripk2
Entrez Gene ID
192656
Gene Name
receptor (TNFRSF)-interacting serine-threonine kinase 2
Gene Aliases
2210420D18Rik, CARD3, CARDIAK, CCK, D4Bwg0615e, RICK, RIP2
Chromosome Location
Chr. 4: 16122741 - 16163679 on Build GRCm38
UniGene ID
Mm.112765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138952.3 NP_620402.1 8 1094
GenBank mRNA AF461040.1 - 8 992
AF487539.1 - 8 992
AK159593.1 - 8 1118
AK172319.1 - 8 1116
AK172417.1 - 8 1116
BC025611.1 - 6 612
BC050845.1 - 8 1108
BC058088.1 - 9 1363
BC069878.1 - 8 1119 5316066

View Full Product Details