Gene Transcripts

Gene Symbol
HHLA3
Entrez Gene ID
11147
Gene Name
HERV-H LTR-associating 3
Gene Aliases
Chromosome Location
Chr. 1: 70353840 - 70368023 on Build GRCh38
UniGene ID
Hs.142245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031693.2 NP_001026863.1 3 770
NM_001036645.1 NP_001031722.1 3 867
NM_001036646.1 NP_001031723.1 3 766
NR_027404.1 4 818
GenBank mRNA AF126163.1 - 3 688
AF126164.1 - 4 799
BC010922.1 - 3 748 4064190

View Full Product Details