Gene Transcripts
- Gene Symbol
- ITGAE
- Entrez Gene ID
- 3682
- Gene Name
- integrin subunit alpha E
- Gene Aliases
- CD103, HUMINAE
- Chromosome Location
- Chr. 17: 3714625 - 3803766 on Build GRCh38
- UniGene ID
- Hs.513867
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002208.4 |
NP_002199.3
|
29, 30 |
3428 |
|
XM_011523823.2 |
XP_011522125.1
|
29, 30 |
3929 |
|
XM_011523825.2 |
XP_011522127.1
|
28, 29 |
3831 |
|
XM_011523827.2 |
XP_011522129.1
|
28, 29 |
3759 |
|
XM_011523828.2 |
XP_011522130.1
|
27, 28 |
3685 |
|
XM_017024586.1 |
XP_016880075.1
|
29, 30 |
5236 |
|
GenBank mRNA |
AF289864.1 |
- |
2, 3 |
242 |
|
BC113436.1 |
- |
29, 30 |
3418 |
8322488 |
BC117207.1 |
- |
29, 30 |
3418 |
40125758 |
L25851.2 |
- |
29, 30 |
3454 |
|
View Full Product Details