Gene Transcripts

Gene Symbol
TEK
Entrez Gene ID
7010
Gene Name
TEK receptor tyrosine kinase
Gene Aliases
CD202B, TIE-2, TIE2, VMCM, VMCM1
Chromosome Location
Chr. 9: 27109141 - 27230178 on Build GRCh38
UniGene ID
Hs.89640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000459.4 NP_000450.2 2 531
NM_001290077.1 NP_001277006.1 2 531
XM_005251561.1 XP_005251618.1 2 198
XM_005251563.1 XP_005251620.1 2 198
GenBank mRNA AB086825.1 - 2 187
AB208796.1 - 2 209
AK291775.1 - 2 219
AK295043.1 - 2 257
BC035514.1 - 2 206 5228999
CA392256.1 - 2 434
DC324016.1 - 2 458
EU826591.1 - 2 81
EU826592.1 - 2 81
L06139.1 - 2 229

View Full Product Details