Gene Transcripts

Gene Symbol
HIGD1A
Entrez Gene ID
25994
Gene Name
HIG1 hypoxia inducible domain family member 1A
Gene Aliases
HIG1, RCF1a
Chromosome Location
Chr. 3: 42782908 - 42804535 on Build GRCh38
UniGene ID
Hs.593134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099668.1 NP_001093138.1 4 1047
NM_001099669.1 NP_001093139.1 4 900
NM_014056.3 NP_054775.2 4 876
GenBank mRNA AF145385.1 - 4 815
AK225571.1 - 4 831
AL110233.1 - 4 846
BC000601.1 - 4 814 3346355
BC009583.1 - 4 807 4093421
BC009594.1 - 4 813 4109190
BC070277.1 - 4 829 4686431

View Full Product Details