Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160036.1 | NP_001153508.1 | 5 | 624 | |
NM_001160037.1 | NP_001153509.1 | 3 | 320 | ||
NM_015178.2 | NP_055993.2 | 3 | 812 | ||
XM_017013251.1 | XP_016868740.1 | 4 | 5139 | ||
GenBank mRNA | AB018260.2 | - | 3 | 812 | |
AK098744.1 | - | 3 | 520 | ||
AK292863.1 | - | 3 | 460 | ||
AK314366.1 | - | 3 | 575 | ||
AY009093.1 | - | 3 | 466 | ||
BC034917.1 | - | 5 | 624 | 4647716 | |
DA289763.1 | - | 3 | 321 |