Gene Transcripts

Gene Symbol
Rbm28
Entrez Gene ID
68272
Gene Name
RNA binding motif protein 28
Gene Aliases
2810480G15Rik, AI503051
Chromosome Location
Chr. 6: 29123573 - 29164724 on Build GRCm38
UniGene ID
Mm.40802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133925.2 NP_598686.2 16 1839
GenBank mRNA AK156773.1 - 16 1776
AK161670.1 - 16 1806
AK167050.1 - 16 1841
BC018373.1 - 12 1213 3708613
BC023293.1 - 6 588
BC024602.1 - 16 1720
BC027123.1 - 9 1145 5356457
BC040811.1 - 16 1784 4015830

View Full Product Details