Gene Transcripts

Gene Symbol
SLC39A14
Entrez Gene ID
23516
Gene Name
solute carrier family 39 member 14
Gene Aliases
HMNDYT2, LZT-Hs4, NET34, ZIP14, cig19
Chromosome Location
Chr. 8: 22367249 - 22434129 on Build GRCh38
UniGene ID
Hs.491232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128431.2 NP_001121903.1 2 318
NM_001135153.1 NP_001128625.1 2 359
NM_001135154.1 NP_001128626.1 2 318
NM_015359.4 NP_056174.2 2 318
XM_005273465.2 XP_005273522.1 3 406
XM_005273466.4 XP_005273523.1 2 390
XM_006716323.2 XP_006716386.1 4 604
XM_006716324.2 XP_006716387.1 3 395
XM_011544478.2 XP_011542780.1 3 470
XM_017013292.1 XP_016868781.1 3 486
XM_017013293.1 XP_016868782.1 4 604
XM_017013294.1 XP_016868783.1 4 513
XM_017013295.1 XP_016868784.1 2 523
GenBank mRNA AK172810.1 - 2 289
AK295807.1 - 2 234
BC015770.1 - 2 280 4865469
D31887.1 - 2 262
DC323208.1 - 2 318
DC423882.1 - 2 360
KU178568.1 - 1 143
KU178569.1 - 1 143
KU178570.1 - 1 143
KU178571.1 - 1 143

View Full Product Details