Gene Transcripts

Gene Symbol
OLA1
Entrez Gene ID
29789
Gene Name
Obg-like ATPase 1
Gene Aliases
DOC45, GBP45, GTBP9, GTPBP9, PTD004
Chromosome Location
Chr. 2: 174072447 - 174248637 on Build GRCh38
UniGene ID
Hs.157351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF078859.1 - 7 794
AF078868.1 - 7 775
AF116703.2 - 4 352
AF134478.1 - 7 750
AK027523.1 - 7 827
AK074710.1 - 6 725
AK225112.1 - 7 763
AK225138.1 - 7 803
AK296339.1 - 7 743
AL136546.1 - 6 682
AL833441.1 - 7 791
BC012842.1 - 7 864 3861915
BC013925.1 - 7 706 3685016
BC029376.1 - 7 743 4658276
BC091522.1 - 7 718 30513213
DQ250006.1 - 7 719

View Full Product Details