Gene Transcripts

Gene Symbol
FEZF2
Entrez Gene ID
55079
Gene Name
FEZ family zinc finger 2
Gene Aliases
FEZ, FEZL, FKSG36, TOF, ZFP312, ZNF312
Chromosome Location
Chr. 3: 62369672 - 62373515 on Build GRCh38
UniGene ID
Hs.241523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018008.3 NP_060478.3 5 1586
GenBank mRNA AF064845.1 - 5 1426
AF332890.1 - 4 1438
AK001004.1 - 3 1121
AK290464.1 - 5 1586
BC022464.1 - 5 1417 4794121

View Full Product Details