Gene Transcripts

Gene Symbol
TENM2
Entrez Gene ID
57451
Gene Name
teneurin transmembrane protein 2
Gene Aliases
ODZ2, TEN-M2, TNM2, ten-2
Chromosome Location
Chr. 5: 166979066 - 168264159 on Build GRCh38
UniGene ID
Hs.631957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080428.2 NP_001073897.2 12, 13 2330
NM_001122679.1 NP_001116151.1 15, 16 2881
XM_005265950.2 XP_005266007.1 17, 18 3528
XM_005265952.2 XP_005266009.1 13, 14 2255
XM_006714897.2 XP_006714960.1 17, 18 3528
XM_011534604.2 XP_011532906.1 16, 17 3342
XM_017009660.1 XP_016865149.1 17, 18 3555
XM_017009661.1 XP_016865150.1 17, 18 3555
XM_017009662.1 XP_016865151.1 16, 17 3345
XM_017009663.1 XP_016865152.1 14, 15 2750
XM_017009664.1 XP_016865153.1 14, 15 2663
XM_017009665.1 XP_016865154.1 14, 15 2459
XM_017009666.1 XP_016865155.1 14, 15 2459
XM_017009667.1 XP_016865156.1 14, 15 2432
XM_017009668.1 XP_016865157.1 14, 15 3969
XM_017009669.1 XP_016865158.1 14, 15 3801
GenBank mRNA AB032953.2 - 6, 7 1018
BX648178.1 - 11, 12 1924
CR936703.1 - 12, 13 2263

View Full Product Details