Gene Transcripts

Gene Symbol
NPVF
Entrez Gene ID
64111
Gene Name
neuropeptide VF precursor
Gene Aliases
C7orf9, RFRP
Chromosome Location
Chr. 7: 25224570 - 25228486 on Build GRCh38
UniGene ID
Hs.60473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022150.3 NP_071433.3 1 105
GenBank mRNA AB040290.1 - 1 58
AF330057.1 - 1 58
AF440392.1 - 1 105
BM714303.1 - 1 50

View Full Product Details