Gene Transcripts

Gene Symbol
ZNF696
Entrez Gene ID
79943
Gene Name
zinc finger protein 696
Gene Aliases
Chromosome Location
Chr. 8: 143291389 - 143299952 on Build GRCh38
UniGene ID
Hs.512740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030895.2 NP_112157.2 3 2339
GenBank mRNA AK024191.1 - 3 2302

View Full Product Details