Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198179.2 | NP_937822.2 | 4 | 1077 | |
GenBank mRNA | AB109629.1 | - | 4 | 698 | |
BC128132.1 | - | 5 | 829 | ||
BC128133.1 | - | 4 | 698 | 40112420 | |
JF810892.1 | - | 4 | 698 |