Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr. 1: 39955112 - 39969968 on Build GRCh38
UniGene ID
Hs.655177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136493.2 NP_001129965.1 7 972
NM_001287808.1 NP_001274737.1 5 673
NM_001287809.1 NP_001274738.1 7 816
NM_032793.4 NP_116182.2 7 933
NR_109896.1 7 933
XM_005271285.1 XP_005271342.1 7 897
XM_011542312.2 XP_011540614.1 7 900
GenBank mRNA AB073383.1 - 7 897
AF289609.1 - 7 889
AF370364.1 - 4 1398
AK074186.1 - 7 838
AK075183.1 - 7 900
AK093223.1 - 7 934
AK291540.1 - 7 895
AK297991.1 - 7 780
AK301533.1 - 5 640
AL834184.1 - 7 887
AY358636.1 - 7 915
BC006353.2 - 3 268
BC011587.1 - 7 907 4646430
BC092414.1 - 7 875 30387073

View Full Product Details