Gene Transcripts
- Gene Symbol
- DMWD
- Entrez Gene ID
- 1762
- Gene Name
- dystrophia myotonica, WD repeat containing
- Gene Aliases
- D19S593E, DMR-N9, DMRN9, gene59
- Chromosome Location
- Chr. 19: 45783006 - 45792802 on Build GRCh38
- UniGene ID
- Hs.515474
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004943.1 |
NP_004934.1
|
2 |
630 |
|
GenBank mRNA |
L19267.1 |
- |
2 |
218 |
|
View Full Product Details