Gene Transcripts

Gene Symbol
DRAM2
Entrez Gene ID
128338
Gene Name
DNA damage regulated autophagy modulator 2
Gene Aliases
CORD21, PRO180, TMEM77, WWFQ154
Chromosome Location
Chr. 1: 111117332 - 111140216 on Build GRCh38
UniGene ID
Hs.485606
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178454.4 NP_848549.3 3 379
XM_005270469.2 XP_005270526.1 4 437
XM_005270470.1 XP_005270527.1 4 560
XM_006710361.1 XP_006710424.1 4 509
XM_006710362.1 XP_006710425.1 4 482
XM_011540707.1 XP_011539009.1 4 378
XM_011540708.1 XP_011539010.1 4 458
XM_017000320.1 XP_016855809.1 3 433
XM_017000321.1 XP_016855810.1 3 292
GenBank mRNA AK309557.1 - 3 616
AL833098.1 - 3 747
AY336747.1 - 3 257
AY358492.1 - 3 242
BC047025.1 - 2 194 5491665
BC091509.1 - 4 471 30513000
DC339928.1 - 3 379
EF710624.1 - 1 122

View Full Product Details