Gene Transcripts

Gene Symbol
MYRFL
Entrez Gene ID
196446
Gene Name
myelin regulatory factor-like
Gene Aliases
C12orf15, C12orf28, bcm1377
Chromosome Location
Chr. 12: 69824518 - 69958726 on Build GRCh38
UniGene ID
Hs.253773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182530.2 NP_872336.2 25 2887
XM_011539052.2 XP_011537354.1 24 3368
XM_011539055.1 XP_011537357.1 23 2355
XM_017018967.1 XP_016874456.1 24 3169
XM_017018968.1 XP_016874457.1 25 3623
XM_017018969.1 XP_016874458.1 24 3103
XM_017018970.1 XP_016874459.1 23 2913
XM_017018973.1 XP_016874462.1 21 2584
GenBank mRNA AK057785.1 - 12 1004
BC101820.1 - 10 839
BC143553.1 - 9 764 9052064
BG187742.1 - 1 62

View Full Product Details