Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000121.3 | NP_000112.1 | 7 | 1027 | |
NR_033663.1 | 7 | 1277 | |||
GenBank mRNA | AK315097.1 | - | 7 | 995 | |
BC112153.1 | - | 7 | 969 | 8327621 | |
BC143697.1 | - | 8 | 1105 | ||
M34986.1 | - | 7 | 892 | ||
M60459.1 | - | 7 | 997 |