Gene Transcripts

Gene Symbol
PRSS33
Entrez Gene ID
260429
Gene Name
protease, serine 33
Gene Aliases
EOS
Chromosome Location
Chr. 16: 2783953 - 2787759 on Build GRCh38
UniGene ID
Hs.280658
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152891.2 NP_690851.2 6 1392
XM_011522451.2 XP_011520753.1 7 1516
XM_011522452.2 XP_011520754.1 7 1328
GenBank mRNA AF536382.1 - 6 1299
BC036846.1 - 7 1374 5245847
BC062334.1 - 6 1394 6110198

View Full Product Details