Gene Transcripts

Gene Symbol
NEK8
Entrez Gene ID
284086
Gene Name
NIMA related kinase 8
Gene Aliases
JCK, NEK12A, NPHP9, RHPD2
Chromosome Location
Chr. 17: 28727899 - 28743457 on Build GRCh38
UniGene ID
Hs.448468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178170.2 NP_835464.1 5 664
XM_011524638.2 XP_011522940.1 6 868
XM_011524640.2 XP_011522942.1 6 868
XM_017024499.1 XP_016879988.1 5 674
XM_017024500.1 XP_016879989.1 5 710
XM_017024501.1 XP_016879990.1 6 868
GenBank mRNA AK301429.1 - 5 674
AY242354.1 - 5 664
AY267371.1 - 5 664
BC112240.1 - 5 698 8327708
BC113705.1 - 5 698 8322757

View Full Product Details