Gene Transcripts

Gene Symbol
GPR150
Entrez Gene ID
285601
Gene Name
G protein-coupled receptor 150
Gene Aliases
PGR11
Chromosome Location
Chr. 5: 95620276 - 95621580 on Build GRCh38
UniGene ID
Hs.143315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199243.1 NP_954713.1 1 52
GenBank mRNA BC030197.1 - 1 250
JF810884.1 - 1 52

View Full Product Details