Gene Transcripts

Gene Symbol
HNRNPCL1
Entrez Gene ID
343069
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 1
Gene Aliases
HNRPCL1
Chromosome Location
Chr. 1: 12847377 - 12848756 on Build GRCh38
UniGene ID
Hs.502617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013631.2 NP_001013653.1 2 787
GenBank mRNA BC002696.2 - 2 756
BC137258.1 - 2 787 9021255
BC171866.1 - 1 625 9054520

View Full Product Details