Gene Transcripts

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
Chromosome Location
Chr. X: 80669488 - 80809736 on Build GRCh38
UniGene ID
Hs.147027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153252.4 NP_694984.4 15 1775
XM_005262113.3 XP_005262170.1 15 1777
XM_017029384.1 XP_016884873.1 11 1305
XM_017029385.1 XP_016884874.1 15 1777
GenBank mRNA AY497046.1 - 15 1775
AY497047.1 - 12 1707
AY497048.1 - 15 1798
AY497049.1 - 14 1768
AY497050.1 - 15 1748
AY497051.1 - 12 1335
AY497052.1 - 17 1934
AY497053.1 - 13 1523
AY497054.1 - 12 1424
AY497055.1 - 15 1709
AY497056.1 - 12 1680
AY497057.1 - 11 1579
AY497058.1 - 11 1418
AY497059.1 - 13 1789
AY497060.1 - 9 1356
BC111490.1 - 15 1725

View Full Product Details