Gene Transcripts

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
Chromosome Location
Chr. X: 80669488 - 80809736 on Build GRCh38
UniGene ID
Hs.147027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153252.4 NP_694984.4 17 2087
XM_005262113.3 XP_005262170.1 17 2089
XM_017029384.1 XP_016884873.1 13 1617
XM_017029385.1 XP_016884874.1 17 2089
GenBank mRNA AY497046.1 - 17 2087
AY497047.1 - 14 2019
AY497048.1 - 17 2110
AY497049.1 - 16 2080
AY497050.1 - 17 2060
AY497051.1 - 14 1647
AY497052.1 - 19 2246
AY497053.1 - 15 1835
AY497054.1 - 14 1736
AY497055.1 - 17 2021
AY497056.1 - 14 1992
AY497057.1 - 13 1891
AY497058.1 - 13 1730
AY497059.1 - 15 2101
AY497060.1 - 11 1668
BC111490.1 - 17 2037

View Full Product Details