Gene Transcripts
- Gene Symbol
- Phb2
- Entrez Gene ID
- 12034
- Gene Name
- prohibitin 2
- Gene Aliases
- AU044498, BAP, Bap37, Bcap37, REA
- Chromosome Location
- Chr. 6: 124712289 - 124716945 on Build GRCm38
- UniGene ID
- Mm.36241
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007531.2 |
NP_031557.2
|
4, 5 |
634 |
|
GenBank mRNA |
AK002334.1 |
- |
4, 5 |
600 |
|
AK011107.1 |
- |
4, 5 |
601 |
|
AK090088.1 |
- |
4, 5 |
590 |
|
AK132052.1 |
- |
4, 5 |
596 |
|
AK132058.1 |
- |
4, 5 |
623 |
|
AK135729.1 |
- |
4, 5 |
552 |
|
AK151952.1 |
- |
4, 5 |
628 |
|
AK152375.1 |
- |
4 |
1283 |
|
AK153368.1 |
- |
4 |
1283 |
|
AK216410.1 |
- |
1 |
18 |
|
AK218588.1 |
- |
1 |
18 |
|
AK219005.1 |
- |
1 |
18 |
|
AY211613.1 |
- |
4, 5 |
596 |
|
AY319418.1 |
- |
4, 5 |
486 |
|
BC145875.1 |
- |
4, 5 |
586 |
40131178 |
X78683.1 |
- |
4, 5 |
590 |
|
View Full Product Details