Gene Transcripts

Gene Symbol
Dio1
Entrez Gene ID
13370
Gene Name
deiodinase, iodothyronine, type I
Gene Aliases
5DI, D1, ITDI1, TXDI1
Chromosome Location
Chr. 4: 107291465 - 107307143 on Build GRCm38
UniGene ID
Mm.148342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007860.3 NP_031886.3 1 136
GenBank mRNA AK002549.1 - 1 137
BC019786.1 - 1 161 5133568
BC037080.1 - 1 172
BC089608.1 - 1 136 6766076
U49861.1 - 1 136

View Full Product Details