Gene Transcripts

Gene Symbol
Itga3
Entrez Gene ID
16400
Gene Name
integrin alpha 3
Gene Aliases
AA407068, CD49C, GAPB3
Chromosome Location
Chr. 11: 95044475 - 95076757 on Build GRCm38
UniGene ID
Mm.57035
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC053031.1 - 12, 13 2049
BC062205.1 - 11, 12 1824 4950774
BC096741.1 - 13, 14 2027
D13867.1 - 12, 13 1671

View Full Product Details