Gene Transcripts

Gene Symbol
GABRA1
Entrez Gene ID
2554
Gene Name
gamma-aminobutyric acid type A receptor alpha1 subunit
Gene Aliases
ECA4, EIEE19, EJM, EJM5
Chromosome Location
Chr. 5: 161847191 - 161899959 on Build GRCh38
UniGene ID
Hs.175934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000806.5 NP_000797.2 8 1158
NM_001127643.1 NP_001121115.1 8 1045
NM_001127644.1 NP_001121116.1 7 1194
NM_001127645.1 NP_001121117.1 7 782
NM_001127648.1 NP_001121120.1 6 804
GenBank mRNA AK223447.1 - 7 931
AK289792.1 - 8 1158
AK295160.1 - 8 977
BC030696.1 - 7 759 4820972
DA232066.1 - 3 454
X13584.1 - 8 911
X14766.1 - 7 904

View Full Product Details