Gene Transcripts

Gene Symbol
Tarbp2
Entrez Gene ID
21357
Gene Name
TAR (HIV) RNA binding protein 2
Gene Aliases
Prbp, TRBP
Chromosome Location
Chr. 15: 102518192 - 102523676 on Build GRCm38
UniGene ID
Mm.291485
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253795.1 NP_001240724.1 4 655
NM_009319.3 NP_033345.2 6 928
GenBank mRNA AK143188.1 - 5 827
AK144205.1 - 4 493
BC002028.1 - 6 641 3489376
BC053432.1 - 2 1103
BQ948575.1 - 6 669
U79962.1 - 6 661

View Full Product Details