Gene Transcripts

Gene Symbol
Pfpl
Entrez Gene ID
56093
Gene Name
pore forming protein-like
Gene Aliases
Epcs5, Epcs50
Chromosome Location
Chr. 19: 12427905 - 12432110 on Build GRCm38
UniGene ID
Mm.285849
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019540.2 NP_062413.2 2 543
GenBank mRNA AF250839.1 - 2 519
AK141262.1 - 2 477
BC052215.1 - 2 511 30020114

View Full Product Details