Gene Transcripts

Gene Symbol
Rev1
Entrez Gene ID
56210
Gene Name
REV1 homolog (S. cerevisiae)
Gene Aliases
1110027I23Rik, AU022044, Rev1l
Chromosome Location
Chr. 1: 38052786 - 38129919 on Build GRCm38
UniGene ID
Mm.389103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019570.3 NP_062516.2 17 2915
GenBank mRNA AB057418.1 - 16 2704
AF179302.1 - 16 2704
AK048838.1 - 17 2919
BC006721.1 - 12 2239
BC058093.1 - 17 2915 6827108

View Full Product Details