Gene Transcripts

Gene Symbol
Rps6kb2
Entrez Gene ID
58988
Gene Name
ribosomal protein S6 kinase, polypeptide 2
Gene Aliases
70kDa, S6K2
Chromosome Location
Chr. 19: 4156792 - 4163383 on Build GRCm38
UniGene ID
Mm.271937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021485.2 NP_067460.1 10 829
GenBank mRNA AJ007938.1 - 10 829
AK014412.1 - 10 908
AK090093.1 - 884
AK156361.1 - 10 882
BC005694.1 - 2 160
BC051631.1 - 10 1135
BC066842.1 - 9 1011 30468251

View Full Product Details