Gene Transcripts

Gene Symbol
Fam212a
Entrez Gene ID
68176
Gene Name
family with sequence similarity 212, member A
Gene Aliases
6230427J02Rik, Inka1
Chromosome Location
Chr. 9: 107984223 - 107985916 on Build GRCm38
UniGene ID
Mm.76166
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026597.3 NP_080873.2 2 575
GenBank mRNA AK020090.1 - 2 539
AK076092.1 - 2 539
BC051538.1 - 1 316
BC115538.1 - 2 514 40101503

View Full Product Details