Gene Transcripts
- Gene Symbol
- Als2cr11
- Entrez Gene ID
- 73463
- Gene Name
- amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human)
- Gene Aliases
- 1700052H20Rik, 4930408G06Rik
- Chromosome Location
- Chr. 1: 59050506 - 59095015 on Build GRCm38
- UniGene ID
- Mm.252811
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015211.1 |
- |
|
530 |
|
AK019575.1 |
- |
|
640 |
|
View Full Product Details