Gene Transcripts

Gene Symbol
Pdzph1
Entrez Gene ID
69239
Gene Name
PDZ and pleckstrin homology domains 1
Gene Aliases
2610034M16Rik, AV114557
Chromosome Location
Chr. 17: 58878808 - 58991375 on Build GRCm38
UniGene ID
Mm.246732
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027001.1 NP_081277.1 3 888
GenBank mRNA BC033350.1 - 3 888
BC034642.1 - 3 848 5363521

View Full Product Details